Assay Details
| Gene Symbol: HNRNPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045911.1 | NP_001039376.1 | 8 - 9 | 967 | 76 | |
| GenBank mRNA | BC105413.1 | - | 8 - 9 | 967 | 76 | |
More Information
Set Membership: |
|

HNRNPA1
Genomic Map