Assay Details
| Gene Symbol: TENT5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045950.1 | NP_001039415.1 | 1 - 2 | 433 | 76 | |
| GenBank mRNA | BC114174.1 | - | 1 - 2 | 433 | 76 | |
More Information
Set Membership: |
|

TENT5B
Genomic Map