Assay Details
| Gene Symbol: ABITRAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045956.1 | NP_001039421.1 | 5 - 6 | 444 | 143 | |
| GenBank mRNA | BC113223.1 | - | 5 - 6 | 444 | 143 | |
More Information
Set Membership: |
|

ABITRAM
Genomic Map