Assay Details
| Gene Symbol: MRPL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045996.2 | NP_001039461.1 | 2 - 3 | 315 | 64 | |
| GenBank mRNA | BC105311.1 | - | 2 - 3 | 316 | 64 | |
More Information
Set Membership: |
|

MRPL28
Genomic Map