Assay Details
Gene Symbol: REM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046001.2 | NP_001039466.1 | 3 - 4 | 698 | 63 | |
GenBank mRNA | BC114085.1 | - | 3 - 4 | 699 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
