Assay Details
Gene Symbol: PXMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046008.2 | NP_001039473.1 | 3 - 4 | 420 | 75 | |
GenBank mRNA | BC112465.1 | - | 3 - 4 | 422 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
