Assay Details
Gene Symbol: ADH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046057.2 | NP_001039522.1 | 3 - 4 | 284 | 69 | |
GenBank mRNA | BC112630.1 | - | 3 - 4 | 285 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
