Assay Details
Gene Symbol: LY6E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046070.1 | NP_001039535.1 | 1 - 2 | 73 | 91 | |
GenBank mRNA | BC114823.1 | - | 1 - 2 | 73 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC114823.1, NM_001046070.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
