Assay Details
| Gene Symbol: EIF2S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046117.2 | NP_001039582.1 | 1 - 2 | 76 | 82 | |
| GenBank mRNA | BC112875.1 | - | 1 - 2 | 77 | 82 | |
| BT030491.1 | - | 1 - 2 | 61 | 82 | ||
More Information
Set Membership: |
|

EIF2S3
Genomic Map