Assay Details
| Gene Symbol: EIF2S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046117.2 | NP_001039582.1 | 2 - 3 | 148 | 97 | |
| GenBank mRNA | BC112875.1 | - | 2 - 3 | 149 | 97 | |
| BT030491.1 | - | 2 - 3 | 133 | 97 | ||
More Information
Set Membership: |
|

EIF2S3
Genomic Map