Assay Details
Gene Symbol: EIF2S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046117.2 | NP_001039582.1 | 4 - 5 | 390 | 60 | |
GenBank mRNA | BC112875.1 | - | 4 - 5 | 391 | 60 | |
BT030491.1 | - | 4 - 5 | 375 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
