Assay Details
Gene Symbol: HSPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046131.2 | NP_001039596.1 | 2 - 3 | 352 | 90 | |
GenBank mRNA | BC105481.1 | - | 2 - 3 | 353 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
