Assay Details
| Gene Symbol: RHOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046138.2 | NP_001039603.1 | 4 - 5 | 456 | 77 | |
| GenBank mRNA | BC114859.1 | - | 4 - 5 | 457 | 77 | |
More Information
Set Membership: |
|

RHOC
Genomic Map