Assay Details
Gene Symbol: PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046144.1 | NP_001039609.1 | 10 - 11 | 1080 | 128 | |
GenBank mRNA | BC113245.1 | - | 10 - 11 | 1080 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
