Assay Details
| Gene Symbol: C14H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046153.3 | NP_001039618.2 | 4 - 5 | 320 | 69 | |
| GenBank mRNA | BC112675.1 | - | 4 - 5 | 304 | 69 | |
| EH210109.1 | - | 4 - 5 | 321 | 69 | ||
More Information
Set Membership: |
|

C14H8orf33
Genomic Map