Assay Details
| Gene Symbol: MDM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046169.1 | NP_001039634.1 | 5 - 6 | 419 | 89 | |
| GenBank mRNA | BC113350.1 | - | 5 - 6 | 419 | 89 | |
More Information
Set Membership: |
|

MDM4
Genomic Map