Assay Details
| Gene Symbol: NAPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046233.2 | NP_001039698.1 | 7 - 8 | 652 | 64 | |
| GenBank mRNA | BC114061.1 | - | 7 - 8 | 655 | 64 | |
More Information
Set Membership: |
|

NAPB
Genomic Map