Assay Details
Gene Symbol: PLPPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046248.2 | NP_001039713.1 | 2 - 3 | 311 | 51 | |
GenBank mRNA | BC114023.1 | - | 2 - 3 | 313 | 51 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC114023.1, NM_001046248.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
