Assay Details
Gene Symbol: HS1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046280.2 | NP_001039745.2 | 2 - 3 | 246 | 88 | |
GenBank mRNA | BC105354.1 | - | 2 - 3 | 249 | 88 | |
EE943328.1 | - | 2 - 3 | 246 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
