Assay Details
| Gene Symbol: C2CD4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046307.1 | NP_001039772.1 | 1 - 2 | 70 | 63 | |
| GenBank mRNA | BC105566.1 | - | 1 - 2 | 70 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105566.1, NM_001046307.1 transcripts.
Set Membership: |
|

C2CD4B
Genomic Map