Assay Details
| Gene Symbol: BAMBI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046309.1 | NP_001039774.1 | 2 - 3 | 666 | 106 | |
| GenBank mRNA | BC114655.1 | - | 2 - 3 | 666 | 106 | |
More Information
Set Membership: |
|

BAMBI
Genomic Map