Assay Details
| Gene Symbol: MRPL44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046321.2 | NP_001039786.1 | 2 - 3 | 663 | 107 | |
| GenBank mRNA | BC112531.1 | - | 2 - 3 | 665 | 107 | |
More Information
Set Membership: |
|

MRPL44
Genomic Map