Assay Details
| Gene Symbol: HSD17B8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046324.1 | NP_001039789.1 | 3 - 4 | 403 | 66 | |
| GenBank mRNA | BC142003.1 | - | 3 - 4 | 401 | 66 | |
| DQ355520.1 | - | 3 - 4 | 403 | 66 | ||
More Information
Set Membership: |
|

HSD17B8
Genomic Map