Assay Details
Gene Symbol: ELAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046327.1 | NP_001039792.1 | 1 - 2 | 162 | 72 | |
GenBank mRNA | BC113356.1 | - | 1 - 2 | 162 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
