Assay Details
Gene Symbol: BPNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046335.1 | NP_001039800.1 | 4 - 5 | 1050 | 86 | |
GenBank mRNA | BC105514.1 | - | 4 - 5 | 1050 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
