Assay Details
| Gene Symbol: SLC10A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046339.1 | NP_001039804.1 | 2 - 3 | 674 | 64 | |
| GenBank mRNA | BC105471.1 | - | 2 - 3 | 674 | 64 | |
| BC146029.1 | - | 2 - 3 | 877 | 64 | ||
More Information
Set Membership: |
|

SLC10A1
Genomic Map