Assay Details
| Gene Symbol: UGT2B10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046354.2 | NP_001039819.1 | 1 - 2 | 760 | 90 | |
| GenBank mRNA | BC112641.1 | - | 1 - 2 | 761 | 90 | |
More Information
Set Membership: |
|

UGT2B10
Genomic Map