Assay Details
Gene Symbol: CNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046379.1 | NP_001039844.1 | 1 - 2 | 126 | 53 | |
GenBank mRNA | BC113327.1 | - | 1 - 2 | 126 | 53 | |
BT030714.1 | - | 1 - 2 | 120 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
