Assay Details
| Gene Symbol: POLR1H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046400.2 | NP_001039865.1 | 2 - 3 | 235 | 84 | |
| GenBank mRNA | BC114799.1 | - | 2 - 3 | 236 | 84 | |
More Information
Set Membership: |
|

POLR1H
Genomic Map