Assay Details
| Gene Symbol: ADCYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046555.1 | NP_001040020.1 | 1 - 2 | 112 | 88 | |
| GenBank mRNA | AY924308.1 | - | 1 - 2 | 112 | 88 | |
| BC123523.1 | - | 2 - 3 | 748 | 88 | ||
More Information
Set Membership: |
|

ADCYAP1
Genomic Map