Assay Details
Gene Symbol: GSTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046560.1 | NP_001040025.1 | 5 - 6 | 403 | 73 | |
GenBank mRNA | BC112491.1 | - | 5 - 6 | 403 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
