Assay Details
| Gene Symbol: CCL25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046569.2 | NP_001040034.2 | 6 - 7 | 1113 | 91 | |
| GenBank mRNA | BC114784.1 | - | 4 - 5 | 433 | 91 | |
| BC118172.1 | - | 6 - 7 | 1113 | 91 | ||
| EU688998.1 | - | 3 - 4 | 340 | 91 | ||
More Information
Set Membership: |
|

CCL25
Genomic Map