Assay Details
| Gene Symbol: NUDT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046608.1 | NP_001040073.1 | 6 - 7 | 1414 | 60 | |
| GenBank mRNA | BC114173.1 | - | 6 - 7 | 1414 | 60 | |
More Information
Set Membership: |
|

NUDT12
Genomic Map