Assay Details
| Gene Symbol: ZNF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046614.1 | NP_001040079.1 | 1 - 2 | 105 | 99 | |
| GenBank mRNA | BC105476.1 | - | 1 - 2 | 105 | 99 | |
| BT025442.1 | - | 1 - 2 | 49 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105476.1, NM_001046614.1 transcripts.
Set Membership: |
|

ZNF3
Genomic Map