Assay Details
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046614.1 | NP_001040079.1 | 2 - 3 | 233 | 103 | |
GenBank mRNA | BC105476.1 | - | 2 - 3 | 233 | 103 | |
BT025442.1 | - | 2 - 3 | 177 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC105476.1, NM_001046614.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
