Assay Details
| Gene Symbol: VPS4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046615.1 | NP_001040080.1 | 9 - 10 | 1256 | 62 | |
| GenBank mRNA | BC113224.1 | - | 9 - 10 | 1256 | 62 | |
More Information
Set Membership: |
|

VPS4A
Genomic Map