Assay Details
Gene Symbol: CCNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075123.1 | NP_001068591.1 | 4 - 5 | 1031 | 61 | |
GenBank mRNA | BC118203.1 | - | 4 - 5 | 1031 | 61 | |
X68321.1 | - | 713 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
