Assay Details
| Gene Symbol: PGRMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075133.1 | NP_001068601.1 | 1 - 2 | 401 | 72 | |
| GenBank mRNA | BC118444.1 | - | 1 - 2 | 401 | 72 | |
| MF084206.1 | - | 1 - 2 | 341 | 72 | ||
More Information
Set Membership: |
|

PGRMC1
Genomic Map