Assay Details
| Gene Symbol: NECTIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075210.2 | NP_001068678.1 | 4 - 5 | 1105 | 51 | |
| GenBank mRNA | BC118434.1 | - | 4 - 5 | 1106 | 51 | |
More Information
Set Membership: |
|

NECTIN2
Genomic Map