Assay Details
| Gene Symbol: PLA2G16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075280.2 | NP_001068748.1 | 3 - 4 | 217 | 95 | |
| GenBank mRNA | BC118454.1 | - | 3 - 4 | 218 | 95 | |
More Information
Set Membership: |
|

PLA2G16
Genomic Map