Assay Details
| Gene Symbol: DUSP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075308.1 | NP_001068776.1 | 2 - 3 | 130 | 66 | |
| GenBank mRNA | BC118267.1 | - | 2 - 3 | 130 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118267.1, NM_001075308.1 transcripts.
Set Membership: |
|

DUSP14
Genomic Map