Assay Details
| Gene Symbol: POP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075312.2 | NP_001068780.1 | 2 - 3 | 199 | 77 | |
| GenBank mRNA | BC116153.1 | - | 2 - 3 | 200 | 77 | |
More Information
Set Membership: |
|

POP5
Genomic Map