Assay Details
| Gene Symbol: DLX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075313.1 | NP_001068781.1 | 2 - 3 | 698 | 86 | |
| GenBank mRNA | BC114749.1 | - | 2 - 3 | 698 | 86 | |
More Information
Set Membership: |
|

DLX5
Genomic Map