Assay Details
Gene Symbol: KDM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075339.1 | NP_001068807.1 | 3 - 4 | 750 | 54 | |
GenBank mRNA | BC123879.1 | - | 3 - 4 | 767 | 54 | |
BT025493.1 | - | 3 - 4 | 750 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
