Assay Details
Gene Symbol: GNPDA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075356.1 | NP_001068824.1 | 5 - 6 | 818 | 154 | |
GenBank mRNA | BC118295.1 | - | 5 - 6 | 818 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
