Assay Details
| Gene Symbol: TOMM40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075438.1 | NP_001068906.1 | 9 - 10 | 1009 | 55 | |
| GenBank mRNA | BC116105.1 | - | 9 - 10 | 1009 | 55 | |
| HM347497.1 | - | 8 - 9 | 953 | 55 | ||
More Information
Set Membership: |
|

TOMM40
Genomic Map