Assay Details
| Gene Symbol: BID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075446.2 | NP_001068914.1 | 5 - 6 | 789 | 60 | |
| GenBank mRNA | BC118359.1 | - | 5 - 6 | 791 | 60 | |
More Information
Set Membership: |
|

BID
Genomic Map