Assay Details
| Gene Symbol: ASF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075453.1 | NP_001068921.1 | 3 - 4 | 517 | 90 | |
| GenBank mRNA | BC118332.1 | - | 3 - 4 | 517 | 90 | |
More Information
Set Membership: |
|

ASF1B
Genomic Map