Assay Details
Gene Symbol: MRPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075474.2 | NP_001068942.1 | 1 - 2 | 182 | 89 | |
GenBank mRNA | BC118470.1 | - | 1 - 2 | 183 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC118470.1, NM_001075474.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
