Assay Details
| Gene Symbol: NT5C3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075515.2 | NP_001068983.2 | 2 - 3 | 164 | 87 | |
| GenBank mRNA | BC118394.1 | - | 2 - 3 | 140 | 87 | |
| EE241538.1 | - | 2 - 3 | 165 | 87 | ||
| EH136301.1 | - | 2 - 3 | 160 | 87 | ||
| EH208684.1 | - | 2 - 3 | 129 | 87 | ||
More Information
Set Membership: |
|

NT5C3B
Genomic Map